missing translation for 'onlineSavingsMsg'
Learn More

3,4,6-Trichloropyridazine 97.0+%, TCI America™

Numéro de catalogue. T34175G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
T34175G 5 g
T34171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. T34175G Fournisseur TCI America Code fournisseur T34175G
Il en reste null

Identifiants chimiques

CAS 6082-66-2
Molecular Formula C4HCl3N2
Molecular Weight (g/mol) 183.42
MDL Number MFCD00834957
InChI Key LJDQXQOPXOLCHL-UHFFFAOYSA-N
PubChem CID 95123
IUPAC Name 3,4,6-trichloropyridazine
SMILES ClC1=CC(Cl)=C(Cl)N=N1

Spécifications

Melting Point 57°C
Color White-Yellow
Boiling Point 98°C
Quantity 5 g
Formula Weight 183.42
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,6-Trichloropyridazine