missing translation for 'onlineSavingsMsg'
Learn More

3,4,6-Tri-O-tert-butyldimethylsilyl-D-glucal, 97%, Thermo Scientific™

Catalog No. AAH6027506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6027506 5 g
AAH6027503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6027506 Supplier Thermo Scientific Chemicals Supplier No. H6027506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Air, water, moisture sensitive..

Chemical Identifiers

CAS 79999-47-6
Molecular Formula C24H52O4Si3
Molecular Weight (g/mol) 488.931
MDL Number MFCD00171721
InChI Key FMXIGEXRLSNRMR-NJYVYQBISA-N
Synonym tri-o-tert-butyldimethylsilyl-d-glucal, 3,4,6-tri-o-t-butyl-dimethylsilyl-d-glucal, 3-o,4-o,6-o-tris tert-butyldimethylsilyl-1,2-dideoxy-d-arabino-1-hexenopyranose, 2r,3r,4r-2-tert-butyldimethylsilyl oxy methyl-3,4-dihydro-2h-pyran-3,4-diyl bis oxy bis tert-butyldimethylsilane, tert-butyl 2r,3r,4r-4-tert-butyldimethylsilyl oxy-2-tert-butyldimethylsilyl oxy methyl-3,4-dihydro-2h-pyran-3-yl oxy dimethylsilane
PubChem CID 10648718
IUPAC Name [(2R,3R,4R)-3,4-bis[[tert-butyl(dimethyl)silyl]oxy]-3,4-dihydro-2H-pyran-2-yl]methoxy-tert-butyl-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)OCC1C(C(C=CO1)O[Si](C)(C)C(C)(C)C)O[Si](C)(C)C(C)(C)C

Specifications

Density 0.928
Boiling Point 210°C (1.2 mmHg)
Flash Point >113°C (235°F)
Refractive Index 1.456
Quantity 5 g
Sensitivity Air and moisture sensitive
Optical Rotation −30° (c=1 in Chloroform)
Formula Weight 488.92
Percent Purity 97%
Chemical Name or Material 3,4,6-Tri-O-tert-butyldimethylsilyl-D-glucal
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen