missing translation for 'onlineSavingsMsg'
Learn More

3,4,6-Tri-O-benzyl-D-glucal, 97%

Catalog No. AAH6043706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6043706 5 g
AAH6043703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6043706 Supplier Thermo Scientific Chemicals Supplier No. H6043706

CAS: 55628-54-1 | C27H28O4 | 416.517 g/mol

3,4,6-Tri-O-benzyl-D-glucal, is an important building block for both solid- and solution-phase synthesis of oligosaccharides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,6-Tri-O-benzyl-D-glucal, is an important building block for both solid- and solution-phase synthesis of oligosaccharides.

Solubility
Insoluble in water, easily soluble in CHCl3

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 55628-54-1
Molecular Formula C27H28O4
Molecular Weight (g/mol) 416.517
MDL Number MFCD00061640
InChI Key MXYLLYBWXIUMIT-PFBJBMPXSA-N
Synonym 3,4,6-tri-o-benzyl-d-glucal, tri-o-benzyl-d-glucal, 2r,3s,4r-3,4-bis benzyloxy-2-benzyloxy methyl-3,4-dihydro-2h-pyran, pubchem10548, 1,5-anhydro-3,4,6-tri-o-benzyl-2-deoxy-d-arabinohex-1-enitol, 1,5-anhydro-2-deoxy-d-arabino-hex-1-enitol 3,4,6-tribenzyl ether
PubChem CID 11742644
IUPAC Name (2R,3S,4R)-3,4-bis(phenylmethoxy)-2-(phenylmethoxymethyl)-3,4-dihydro-2H-pyran
SMILES C1=CC=C(C=C1)COCC2C(C(C=CO2)OCC3=CC=CC=C3)OCC4=CC=CC=C4

Specifications

Melting Point 57°C to 58°C
Quantity 5 g
Beilstein 1691968
Sensitivity Hygroscopic
Solubility Information Insoluble in water,easily soluble in CHCl3
Optical Rotation −2.7° (c=5 in Chloroform)
Formula Weight 416.5
Percent Purity 97%
Chemical Name or Material 3,4,6-Tri-O-benzyl-D-glucal
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only