missing translation for 'onlineSavingsMsg'
Learn More

3,4,6-Tri-O-acetyl-D-glucal, 98%, Thermo Scientific Chemicals

Catalog No. AAL1410314
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1410314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1410314 Supplier Thermo Scientific Chemicals Supplier No. L1410314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2873-29-2 | Molecular Formula: C12H16O7 | Molecular Weight: 272.253 g/mol

3,4,6-Tri-O-acetyl-D-glucal acts as a building block for the synthesis of oligosaccharides in both solution and solid-phase. Further, it is used in the preparation of D-arabino-1,5-anhydro-2-deoxy-hex-1-enitol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,6-Tri-O-acetyl-D-glucal acts as a building block for the synthesis of oligosaccharides in both solution and solid-phase. Further, it is used in the preparation of D-arabino-1,5-anhydro-2-deoxy-hex-1-enitol.

Solubility
Soluble in chloroform and ethanol. Insoluble in water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 2873-29-2
Molecular Formula C12H16O7
Molecular Weight (g/mol) 272.253
MDL Number MFCD00063253
InChI Key LLPWGHLVUPBSLP-UTUOFQBUSA-N
Synonym tri-o-acetyl-d-glucal, 3,4,6-tri-o-acetyl-d-glucal, 2r,3s,4r-2-acetoxymethyl-3,4-dihydro-2h-pyran-3,4-diyl diacetate, d-glucal triacetate, 3,4,6-tri-o-acetyl-1,5-anhydro-2-deoxy-d-arabino-hex-1-enitol, arabino-hex-1-enitol, 1,5-anhydro-2-deoxy-, triacetate, tri-o-acetylglucal, triacetyl-d-glucal, 2r,3s,4r-3,4-bis acetyloxy-3,4-dihydro-2h-pyran-2-yl methyl acetate, tri-o-acetyl-glucal
PubChem CID 688303
IUPAC Name [(2R,3S,4R)-3,4-diacetyloxy-3,4-dihydro-2H-pyran-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C=CO1)OC(=O)C)OC(=O)C

Specifications

Melting Point 52°C to 55°C
Quantity 25 g
Beilstein 90781
Solubility Information Soluble in chloroform and ethanol. Insoluble in water.
Optical Rotation −25° (c=1 in Methanol)
Formula Weight 272.26
Percent Purity 98%
Chemical Name or Material 3,4,6-Tri-O-acetyl-D-glucal
EINECSNumber 220-709-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only