missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxyphenol, 97%

Catalog No. AAL0152714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0152714 25 g
AAL0152706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0152714 Supplier Thermo Scientific Chemicals Supplier No. L0152714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 642-71-7 | C9H12O4 | 184.191 g/mol

3,4,5-Trimethoxyphenol is used as Flavonoid drug intermediates. It is used for pharmaceutical, cosmetics, agrochemicals and food industry research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,5-Trimethoxyphenol is used as Flavonoid drug intermediates. It is used for pharmaceutical, cosmetics, agrochemicals and food industry research.

Solubility
Soluble in chloroform.

Notes
Light Sensitive. Store in the dark. Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances.

Chemical Identifiers

CAS 642-71-7
Molecular Formula C9H12O4
Molecular Weight (g/mol) 184.191
MDL Number MFCD00008389
InChI Key VTCDZPUMZAZMSB-UHFFFAOYSA-N
Synonym antiarol, phenol, 3,4,5-trimethoxy, 3,4,5-trimethoxy phenol, spectrum_000525, specplus_000623, spectrum3_001208, spectrum4_001487, spectrum5_000193, 3,4,5-trirnethoxyphenol, 3,4,5-trimethoxy-phenol
PubChem CID 69505
ChEBI CHEBI:2760
IUPAC Name 3,4,5-trimethoxyphenol
SMILES COC1=CC(=CC(=C1OC)OC)O

Specifications

Melting Point 143°C to 148°C
Quantity 25 g
Beilstein 1958573
Sensitivity Light sensitive
Solubility Information Soluble in chloroform.
Formula Weight 184.19
Percent Purity 97%
Chemical Name or Material 3,4,5-Trimethoxyphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-387-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only