missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4,5-Trimethoxybenzoic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 170°C |
| Color | White to Yellow |
| Boiling Point | 227°C |
| Quantity | 500 g |
| Formula Weight | 212.2 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3,4,5-Trimethoxybenzoic Acid |
Safety and Handling
| EINECSNumber | (3)-1627 |
| TSCA | Yes |