missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4,5-Trimethoxybenzoic acid, 99+%
Description
3,4,5-Trimethoxybenzoic acid is used as intermediates in medicine and organic synthesis. It acts as a metabolite of trimebutine. It is widely used in making dyes and inks, photographic developers and used as astringents in medically.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,4,5-Trimethoxybenzoic acid is used as intermediates in medicine and organic synthesis. It acts as a metabolite of trimebutine. It is widely used in making dyes and inks, photographic developers and used as astringents in medically.
Solubility
Soluble in ethanol, ethyl ether and chloroform. Slightly soluble in water.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
3,4,5-Trimethoxybenzoic acid is used as intermediates in medicine and organic synthesis. It acts as a metabolite of trimebutine. It is widely used in making dyes and inks, photographic developers and used as astringents in medically.
Solubility
Soluble in ethanol, ethyl ether and chloroform. Slightly soluble in water.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications
Specifications
| Melting Point | 168°C to 171°C |
| Boiling Point | 225°C to 226°C (10 mmHg) |
| Quantity | 500 g |
| Beilstein | 884655 |
| Solubility Information | Soluble in ethanol,ethyl ether and chloroform. Slightly soluble in water. |
| Formula Weight | 212.2 |
| Percent Purity | ≥99% |
| Chemical Name or Material | 3,4,5-Trimethoxybenzoic acid |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 204-248-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only