missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trifluorobenzonitrile 98.0+%, TCI America™

Catalog No. T23701G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23701G 1 g
T23705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23701G Supplier TCI America Supplier No. T23701G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 134227-45-5
Molecular Formula C7H2F3N
Molecular Weight (g/mol) 157.095
MDL Number MFCD00074948
InChI Key XFKYJMGXZXJYBS-UHFFFAOYSA-N
Synonym 3,4,5-trifluoro-1-cyano benzene, benzonitrile, 3,4,5-trifluoro, 3,4,5-trifluoro-benzonitrile, 3,4,5-trifluorobenzenecarbonitrile, pubchem1573, intermediates-zcf02038, acmc-1bz57, ksc493s4j, 3,4,5-?trifluorobenzonitrile, 3,4,5-trifluorobenzonitrile
PubChem CID 593814
IUPAC Name 3,4,5-trifluorobenzonitrile
SMILES C1=C(C=C(C(=C1F)F)F)C#N

Specifications

Melting Point 47°C
Color White-Yellow
Boiling Point 69°C
Quantity 1 g
UN Number 3439
Formula Weight 157.10
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5-Trifluorobenzonitrile