missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Methylenebis(1-tert-butyl-3-imidazolium Bromide) 98.0+%, TCI America™

Catalog No. M27901G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27901G 1 g
M27905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27901G Supplier TCI America Supplier No. M27901G
Only null left

Chemical Identifiers

CAS 247902-48-3
Molecular Formula C15H26Br2N4
Molecular Weight (g/mol) 422.209
InChI Key WKZMWHMEXVUXBS-UHFFFAOYSA-L
PubChem CID 11112632
IUPAC Name 1-tert-butyl-3-[(3-tert-butylimidazol-1-ium-1-yl)methyl]imidazol-3-ium;dibromide
SMILES CC(C)(C)N1C=C[N+](=C1)C[N+]2=CN(C=C2)C(C)(C)C.[Br-].[Br-]

Specifications

Melting Point 225°C
Quantity 1 g
Formula Weight 422.21
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Methylenebis(1-tert-butyl-3-imidazolium Bromide)