missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dimethyltriphenylamine 98.0+%, TCI America™

Numéro de catalogue. D47551G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D47551G 1 g
D47555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. D47551G Fournisseur TCI America Code fournisseur D47551G
Il en reste null

Identifiants chimiques

CAS 13511-11-0
Molecular Formula C20H19N
Molecular Weight (g/mol) 273.379
MDL Number MFCD23115649
InChI Key ZCAWQFNYHFHEPZ-UHFFFAOYSA-N
Synonym Bis(3-methylphenyl)phenylamine, N,N-Di-m-tolylaniline, Phenyl(di-m-tolyl)amine
PubChem CID 18326012
IUPAC Name 3-methyl-N-(3-methylphenyl)-N-phenylaniline
SMILES CC1=CC(=CC=C1)N(C2=CC=CC=C2)C3=CC=CC(=C3)C

Spécifications

Melting Point 72°C
Color White-Yellow
Quantity 1 g
Formula Weight 273.38
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Dimethyltriphenylamine