missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylbutyryl Chloride 99.0+%, TCI America™

Catalog No. B119625ML
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B119625ML 25 mL
B1196250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B119625ML Supplier TCI America Supplier No. B119625ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7065-46-5
Molecular Formula C6H11ClO
Molecular Weight (g/mol) 134.603
MDL Number MFCD00000737
InChI Key BUTKIHRNYUEGKB-UHFFFAOYSA-N
Synonym 3,3-dimethylbutyryl chloride, tert-butylacetyl chloride, t-butylacetyl chloride, butanoyl chloride, 3,3-dimethyl, 3,3-dimethyl butyryl chloride, tert-butyl acetyl chloride, neohexanoyl chloride, t-butylacetylchloride, tertbutylacetylchloride, pubchem2093
PubChem CID 81514
IUPAC Name 3,3-dimethylbutanoyl chloride
SMILES CC(C)(C)CC(=O)Cl

Specifications

Boiling Point 132°C
Quantity 25 mL
UN Number 2920
Formula Weight 134.60
Percent Purity ≥99.0% (T)
Physical Form Liquid
Chemical Name or Material 3,3-Dimethylbutyryl Chloride
EINECSNumber (2)-0632
TSCA Yes