missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dimethylbiphenyl 98.0+%, TCI America™

Catalog No. p-7126724
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D14305ML 5 mL
D143025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D14305ML Supplier TCI America Supplier No. D14305ML

Chemical Identifiers

CAS 612-75-9
Molecular Formula C14H14
Molecular Weight (g/mol) 182.27
MDL Number MFCD00008534
InChI Key GVEDOIATHPCYGS-UHFFFAOYSA-N
Synonym 3,3'-dimethylbiphenyl, 3,3'-dimethyl-1,1'-biphenyl, 3,3'-ditolyl, m,m'-bitolyl, 1,1'-biphenyl, 3,3'-dimethyl, 1-methyl-3-3'-methylphenyl benzene, 1,1'-biphenyl,3,3'-dimethyl, 1-methyl-3-3-methylphenyl benzene, pubchem3146, 3,3-dimethyldiphenyl
PubChem CID 11931
IUPAC Name 3,3'-dimethyl-1,1'-biphenyl
SMILES CC1=CC(=CC=C1)C1=CC(C)=CC=C1

Specifications

Color Yellow
Boiling Point 155°C
Quantity 5 mL
Formula Weight 182.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,3′-Dimethylbiphenyl