missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3'-Dimethylazobenzene 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 53°C |
| Quantity | 1 g |
| Formula Weight | 210.28 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3,3′-Dimethylazobenzene |
Sécurité et manipulation
| TSCA | No |