missing translation for 'onlineSavingsMsg'
Learn More

3-[3-(Dimethylamino)propoxy]-N-methylbenzylamine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC59446DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC59446DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

910037-03-5
222.33
ODWHICGCLWFLBT-UHFFFAOYSA-N
24229606
CNCC1=CC(OCCCN(C)C)=CC=C1
C13H22N2O
MFCD09064992
3-3-dimethylamino propoxy-n-methylbenzylamine, dimethyl 3-3-methylamino methyl phenoxy propyl amine, 3-3-dimethylamino propoxy phenyl methyl methyl amine, n,n-dimethyl-3-3-methylamino methyl phenoxy propan-1-amine, benzenemethanamine, 3-3-dimethylamino propoxy-n-methyl, benzenemethanamine,3-3-dimethylamino propoxy-n-methyl
N,N-dimethyl-3-[3-(methylaminomethyl)phenoxy]propan-1-amine

Specifications

3-[3-(Dimethylamino)propoxy]-N-methylbenzylamine
94.5
95%
C13H22N2O
MFCD09064992
ODWHICGCLWFLBT-UHFFFAOYSA-N
N,N-dimethyl-3-[3-(methylaminomethyl)phenoxy]propan-1-amine
24229606
95%
910037-03-5
100.0
Amber Glass Bottle
1g
3-3-dimethylamino propoxy-n-methylbenzylamine, dimethyl 3-3-methylamino methyl phenoxy propyl amine, 3-3-dimethylamino propoxy phenyl methyl methyl amine, n,n-dimethyl-3-3-methylamino methyl phenoxy propan-1-amine, benzenemethanamine, 3-3-dimethylamino propoxy-n-methyl, benzenemethanamine,3-3-dimethylamino propoxy-n-methyl
CNCC1=CC(OCCCN(C)C)=CC=C1
222.33
222.33
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents