Learn More
3,3-Dimethyl-1-(trifluoromethyl)-1,2-benziodoxole, 95%, Thermo Scientific™
$400.26 - $1,593.82
Chemical Identifiers
| CAS | 887144-97-0 |
|---|---|
| Molecular Formula | C10H10F3IO |
| Molecular Weight (g/mol) | 330.09 |
| InChI Key | HVAPLSNCVYXFDQ-UHFFFAOYSA-N |
| Synonym | 3,3-dimethyl-1-trifluoromethyl-1,2-benziodoxole, 1-trifluoromethyl-3,3-dimethyl-1,2-benziodoxole, togni's reagent, togni reagent, unii-3ok0e02mt6, 1,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl, 1,3-dihydro-3,3-dimethyl-1-trifluoromethyl-1,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl-1,3-dihydro-1lambda3,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl-1? 3 ,2-benziodaoxole, togni's reagents dimethyl compound mi |
| PubChem CID | 16043572 |
| IUPAC Name | 3,3-dimethyl-1-(trifluoromethyl)-1$l^{3},2-benziodoxole |
| SMILES | CC1(C2=CC=CC=C2I(O1)C(F)(F)F)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC456902500
|
N/A | 250mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC456900010
|
N/A | 1g |
Each for $400.26
|
|
|||||
|
AC456900050
|
N/A | 5g |
Each for $1,593.82
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 887144-97-0 | |
| 330.09 | |
| 3,3-dimethyl-1-trifluoromethyl-1,2-benziodoxole, 1-trifluoromethyl-3,3-dimethyl-1,2-benziodoxole, togni's reagent, togni reagent, unii-3ok0e02mt6, 1,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl, 1,3-dihydro-3,3-dimethyl-1-trifluoromethyl-1,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl-1,3-dihydro-1lambda3,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl-1? 3 ,2-benziodaoxole, togni's reagents dimethyl compound mi | |
| 3,3-dimethyl-1-(trifluoromethyl)-1$l^{3},2-benziodoxole |
| C10H10F3IO | |
| HVAPLSNCVYXFDQ-UHFFFAOYSA-N | |
| 16043572 | |
| CC1(C2=CC=CC=C2I(O1)C(F)(F)F)C |
Specifications
| 887144-97-0 | |
| 94% min. (HPLC) | |
| 3,3-dimethyl-1-trifluoromethyl-1,2-benziodoxole, 1-trifluoromethyl-3,3-dimethyl-1,2-benziodoxole, togni's reagent, togni reagent, unii-3ok0e02mt6, 1,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl, 1,3-dihydro-3,3-dimethyl-1-trifluoromethyl-1,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl-1,3-dihydro-1lambda3,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl-1? 3 ,2-benziodaoxole, togni's reagents dimethyl compound mi | |
| CC1(C2=CC=CC=C2I(O1)C(F)(F)F)C | |
| 330.09 | |
| 330.09 | |
| 3,3-Dimethyl-1-(trifluoromethyl)-1,2-benziodoxole |
| Authentic | |
| C10H10F3IO | |
| HVAPLSNCVYXFDQ-UHFFFAOYSA-N | |
| 3,3-dimethyl-1-(trifluoromethyl)-1$l^{3},2-benziodoxole | |
| 16043572 | |
| 95% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
RUO – Research Use Only