missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyl-1-(trifluoromethyl)-1,2-benziodoxole, 95%

Catalog No. AC456900010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456900010 1 g
AC456900050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC456900010 Supplier Thermo Scientific Chemicals Supplier No. 456900010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 887144-97-0 | C10H10F3IO | 330.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 887144-97-0
Molecular Formula C10H10F3IO
Molecular Weight (g/mol) 330.09
InChI Key HVAPLSNCVYXFDQ-UHFFFAOYSA-N
Synonym 3,3-dimethyl-1-trifluoromethyl-1,2-benziodoxole, 1-trifluoromethyl-3,3-dimethyl-1,2-benziodoxole, togni's reagent, togni reagent, unii-3ok0e02mt6, 1,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl, 1,3-dihydro-3,3-dimethyl-1-trifluoromethyl-1,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl-1,3-dihydro-1lambda3,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl-1? 3 ,2-benziodaoxole, togni's reagents dimethyl compound mi
PubChem CID 16043572
IUPAC Name 3,3-dimethyl-1-(trifluoromethyl)-1$l^{3},2-benziodoxole
SMILES CC1(C2=CC=CC=C2I(O1)C(F)(F)F)C

Specifications

Melting Point 75.0°C to 79.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 330.09
Percent Purity 95%
Chemical Name or Material 3,3-Dimethyl-1-(trifluoromethyl)-1,2-benziodoxole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only