missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyl-1-butene 96.0+%, TCI America™

Catalog No. D116325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D116325ML 25 mL
D1163100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D116325ML Supplier TCI America Supplier No. D116325ML

Chemical Identifiers

CAS 558-37-2
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00008853
InChI Key PKXHXOTZMFCXSH-UHFFFAOYSA-N
Synonym 3,3-dimethyl-1-butene, neohexene, 1-butene, 3,3-dimethyl, tert-butylethylene, tert-butylethene, tert-hexene, trimethylvinylmethane, 3,3-dimethylbutene, 2,2-dimethyl-3-butene, unii-01zb73d2kk
PubChem CID 11210
IUPAC Name 3,3-dimethylbut-1-ene
SMILES CC(C)(C)C=C

Specifications

Color Colorless
Boiling Point 41°C
Quantity 25 mL
UN Number 3295
Formula Weight 84.16
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,3-Dimethyl-1-butene