missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diethylthiacarbocyanine Iodide 98.0+%, TCI America™

Catalog No. p-7128283
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D44541G Supplier TCI America Supplier No. D44541G

Chemical Identifiers

CAS 905-97-5
Molecular Formula C21H22IN2S2+
Molecular Weight (g/mol) 493.445
MDL Number MFCD00011956
InChI Key VZBILKJHDPEENF-UHFFFAOYSA-N
Synonym 3,3'-diethylthiacarbocyanine iodide
PubChem CID 129893628
IUPAC Name (2Z)-3-ethyl-2-[(E)-3-(3-ethyl-1,3-benzothiazol-3-ium-2-yl)prop-2-enylidene]-1,3-benzothiazole;hydroiodide
SMILES CCN1C2=CC=CC=C2SC1=CC=CC3=[N+](C4=CC=CC=C4S3)CC.I

Specifications

Melting Point 268°C
Quantity 1 g
Formula Weight 492.44
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Diethylthiacarbocyanine Iodide
RTECSNumber DL7061500
TSCA No