missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diethyloxadicarbocyanine Iodide 98.0+%, TCI America™

Catalog No. D4457200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4457200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4457200MG Supplier TCI America Supplier No. D4457200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14806-50-9
Molecular Formula C23H23IN2O2
Molecular Weight (g/mol) 486.35
MDL Number MFCD00011953,MFCD00011953
InChI Key CLDZYSUDOQXJOU-UHFFFAOYSA-M
Synonym dodc iodide, 3-ethyl-2-5-3-ethyl-2-benzoxazolinylidene-1,3-pentadienyl benzoxazolium iodide, 5-3-ethylbenzoxazol-2-ylidene-1-3-ethylbenzoxazolium-2-yl-1,3-pentadiene iodide
PubChem CID 91972131
ChEBI CHEBI:51899
IUPAC Name 3-ethyl-2-[(3E)-5-[(2Z)-3-ethyl-2,3-dihydro-1,3-benzoxazol-2-ylidene]penta-1,3-dien-1-yl]-1,3-benzoxazol-3-ium iodide
SMILES [I-].CCN1\C(OC2=CC=CC=C12)=C\C=C\C=CC1=[N+](CC)C2=CC=CC=C2O1

Specifications

Color Blue
Quantity 200 mg
Formula Weight 486.35
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Diethyloxadicarbocyanine Iodide