missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diethyloxacarbocyanine Iodide 98.0+%, TCI America™

Catalog No. D44535G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44535G 5 g
D44531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44535G Supplier TCI America Supplier No. D44535G
Only null left

Chemical Identifiers

CAS 905-96-4
Molecular Formula C21H22IN2O2+
Molecular Weight (g/mol) 461.323
MDL Number MFCD00011952
InChI Key FIZZUEJIOKEFFZ-UHFFFAOYSA-N
Synonym 3,3'-diethyloxacarbocyanine iodide
PubChem CID 129893627
IUPAC Name (2Z)-3-ethyl-2-[(E)-3-(3-ethyl-1,3-benzoxazol-3-ium-2-yl)prop-2-enylidene]-1,3-benzoxazole;hydroiodide
SMILES CCN1C2=CC=CC=C2OC1=CC=CC3=[N+](C4=CC=CC=C4O3)CC.I

Specifications

Melting Point 276°C
Quantity 5 g
Formula Weight 460.32
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Diethyloxacarbocyanine Iodide