missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diaminodiphenylmethane 98.0+%, TCI America™

Numéro de catalogue. D16831G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D16831G 1 g
D16835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. D16831G Fournisseur TCI America Code fournisseur D16831G
Il en reste null

Identifiants chimiques

CAS 19471-12-6
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.269
MDL Number MFCD00014777
InChI Key CKOFBUUFHALZGK-UHFFFAOYSA-N
PubChem CID 146486
IUPAC Name 3-[(3-aminophenyl)methyl]aniline
SMILES C1=CC(=CC(=C1)N)CC2=CC(=CC=C2)N

Spécifications

Melting Point 85°C
Color White-Yellow
Quantity 1 g
Synonym 3,3′C-Methylenedianiline
Formula Weight 198.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Diaminodiphenylmethane