missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Bromophenyl)-5-methyl-1,2,4-oxadiazole, ≥97%, Thermo Scientific™

Numéro de catalogue. p-3174887
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC39110CB 250mg
CC39110DA 1g
CC39110DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. CC39110CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC39110CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 160377-57-1
Formule moléculaire C9H7BrN2O
Poids moléculaire (g/mol) 239.072
Numéro MDL MFCD08087563
Clé InChI GTYLSVIVKRJHIQ-UHFFFAOYSA-N
Synonyme 3-3-bromophenyl-5-methyl-1,2,4-oxadiazole, 1,2,4-oxadiazole, 3-3-bromophenyl-5-methyl, acmc-1c5gb, chembrdg-bb 9035421, 3-3-bromophenyl-5-methyl-1,2,4-oxadiazol, 3-3-bromophenyl-5-methyl-1,2,4 oxadiazole, 3-3-bromophenyl-5-methyl-1,2,4-oxadiazole saltdata: free
PubChem CID 8895139
Nom de l’IUPAC 3-(3-bromophenyl)-5-methyl-1,2,4-oxadiazole
SOURIRES CC1=NC(=NO1)C2=CC(=CC=C2)Br

Spécifications

Nom ou substance chimique 3-(3-Bromophenyl)-5-methyl-1, 2, 4-oxadiazole
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 250mg
Poids de formule 239.07
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.