missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3'-Bipyridyl 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 175°C |
| Quantity | 5 g |
| Synonym | 3,3′-Bipyridine, 3,3′-Dipyridyl |
| Formula Weight | 156.19 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 3,3′-Bipyridyl |
Safety and Handling
| RTECSNumber | DW1758000 |
| TSCA | No |