missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3,5-Trimethylcyclohexyl Salicylate (cis- and trans- mixture) 98.0+%, TCI America™
Supplier: TCI America T2278500G
Specifications
| 3,3,5-Trimethylcyclohexyl Salicylate (cis- and trans- mixture) | |
| Colorless | |
| C16H22O3 | |
| homosalate, coppertone, homomenthyl salicylate, 3,3,5-trimethylcyclohexyl salicylate, heliopan, heliophan, filtersol a, m-homomenthyl salicylate, homosalatum, benzoic acid, 2-hydroxy-, 3,3,5-trimethylcyclohexyl ester | |
| CC1CC(CC(C1)(C)C)OC(=O)C2=CC=CC=C2O | |
| 262.349 | |
| 262.35 | |
| Liquid |
| 118-56-9 | |
| 165°C | |
| 500 g | |
| WSSJONWNBBTCMG-UHFFFAOYSA-N | |
| (3,3,5-trimethylcyclohexyl) 2-hydroxybenzoate | |
| 8362 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 118-56-9 | |
| 262.349 | |
| homosalate, coppertone, homomenthyl salicylate, 3,3,5-trimethylcyclohexyl salicylate, heliopan, heliophan, filtersol a, m-homomenthyl salicylate, homosalatum, benzoic acid, 2-hydroxy-, 3,3,5-trimethylcyclohexyl ester | |
| (3,3,5-trimethylcyclohexyl) 2-hydroxybenzoate |
| C16H22O3 | |
| WSSJONWNBBTCMG-UHFFFAOYSA-N | |
| 8362 | |
| CC1CC(CC(C1)(C)C)OC(=O)C2=CC=CC=C2O |
Safety and Handling
EINECSNumber : (3)-3285
TSCA : Yes