missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetrachlorodiphenyl Disulfide 99.0+%, TCI America™

Catalog No. T181325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T181325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T181325G Supplier TCI America Supplier No. T181325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 137897-99-5
Molecular Formula C12H6Cl4S2
Molecular Weight (g/mol) 356.1
MDL Number MFCD01862261
InChI Key JMQANWHMOHXBEA-UHFFFAOYSA-N
Synonym Bis(3,5-dichlorophenyl) Disulfide
PubChem CID 2735516
IUPAC Name 1,3-dichloro-5-[(3,5-dichlorophenyl)disulfanyl]benzene
SMILES C1=C(C=C(C=C1Cl)Cl)SSC2=CC(=CC(=C2)Cl)Cl

Specifications

Melting Point 67°C
Color Yellow
Quantity 25 g
Formula Weight 356.10
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′,5,5′-Tetrachlorodiphenyl Disulfide
EINECSNumber (3)-4544
TSCA No