missing translation for 'onlineSavingsMsg'
Learn More

3-(3,4-Dihydroxyphenyl)-L-alanine, 99%

Catalog No. AC167531000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167531000 100 g Glass Bottle
AC167530050 5 g Glass Bottle
AC167530250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC167531000 Supplier Thermo Scientific Chemicals Supplier No. 167531000

3-(3,4-Dihydroxyphenyl)-L-alanine (levodopa orL-dopa), CAS # 59-92-7, is a dopamine precursor molecule. | CAS: 59-92-7 | C9H11NO4 | 197.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• 3-(3,4-Dihydroxyphenyl)-L-alanine, or L-dopa, is converted to dopamine by DOPA decarboxylase in the body

• Upon conversion to dopamine, it acts as an excitatory neurotransmitter

Applications

• L-dopa is useful in the laboratory study of Parkinson’s to replenish dopamine in depleted circuits

• L-dopa has also been used for laboratory research on hypertension and melanocyte activity

Chemical Identifiers

CAS 59-92-7
Molecular Formula C9H11NO4
Molecular Weight (g/mol) 197.19
MDL Number MFCD00002598
InChI Key WTDRDQBEARUVNC-LURJTMIESA-N
Synonym levodopa, l-dopa, dopar, 3-hydroxy-l-tyrosine, 3,4-dihydroxy-l-phenylalanine, larodopa, bendopa, levopa, dihydroxy-l-phenylalanine, cidandopa
PubChem CID 6047
ChEBI CHEBI:15765
IUPAC Name (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C(O)=C1)C(O)=O

Specifications

Color Cream to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Loss on Drying 1% max.
Packaging Glass Bottle
Linear Formula (HO)2C6H3CH2CH(NH2)CO2H
Beilstein 14, IV, 2492
Merck Index 15, 5516
Sulfated Ash 0.1% max.
Quantity 100 g
Formula Weight 197.19
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 3-(3, 4-Dihydroxyphenyl)-L-alanine
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only