missing translation for 'onlineSavingsMsg'
Learn More

3,3',4',7-Tetrahydroxyflavone, 96%

Catalog No. AAL14207MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL14207MD 250 mg
AAL1420703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL14207MD Supplier Thermo Scientific Chemicals Supplier No. L14207MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 528-48-3 | C15H10O6 | 286.239 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 528-48-3
Molecular Formula C15H10O6
Molecular Weight (g/mol) 286.239
MDL Number MFCD00006829
InChI Key XHEFDIBZLJXQHF-UHFFFAOYSA-N
Synonym fisetin, viset, 5-desoxyquercetin, fustel, 2-3,4-dihydroxyphenyl-3,7-dihydroxy-4h-chromen-4-one, fisetholz, superfustel, cotinin, fietin, fustet
PubChem CID 5281614
ChEBI CHEBI:42567
IUPAC Name 2-(3,4-dihydroxyphenyl)-3,7-dihydroxychromen-4-one
SMILES C1=CC(=C(C=C1C2=C(C(=O)C3=C(O2)C=C(C=C3)O)O)O)O

Specifications

Melting Point 330°C (decomposition)
Quantity 250 mg
Beilstein 292829
Merck Index 14,4088
Formula Weight 286.24
Percent Purity 97%
Chemical Name or Material 3,3',4',7-Tetrahydroxyflavone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-434-4
RTECSNumber LK9250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only