missing translation for 'onlineSavingsMsg'
Learn More

3,3,3-Trifluoropropionic Acid 98.0+%, TCI America™

Catalog No. T17135G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17135G 5 g
T17131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17135G Supplier TCI America Supplier No. T17135G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2516-99-6
Molecular Formula C3H3F3O2
Molecular Weight (g/mol) 128.05
MDL Number MFCD00153292
InChI Key KSNKQSPJFRQSEI-UHFFFAOYSA-N
Synonym 3,3,3-trifluoropropionic acid, 3,3,3-trifluoro-propionic acid, propanoic acid, 3,3,3-trifluoro, tfpa, acmc-209ghu, trifluoromethylacetic acid, trifluoromethyl acetic acid, ksc203o6p, 3,3,3,-trifluoropropionic acid, 3,3,3-tri-fluoropropionic acid
PubChem CID 2777972
IUPAC Name 3,3,3-trifluoropropanoic acid
SMILES C(C(=O)O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 153°C
Quantity 5 g
UN Number 2924
Formula Weight 128.05
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3,3,3-Trifluoropropionic Acid