missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3(2H)-Pyridazinone 96.0+%, TCI America™
Specifications
Specifications
| Melting Point | 102°C |
| Color | White-Yellow |
| Boiling Point | 101°C |
| Quantity | 5 g |
| Formula Weight | 96.09 |
| Percent Purity | ≥96.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3(2H)-Pyridazinone |
Safety and Handling
| TSCA | No |