missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Thienyl)propanoic acid, 98%

Catalog No. AC330110010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC330110010 1 g Glass bottle
AC330110050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC330110010 Supplier Thermo Scientific Chemicals Supplier No. 330110010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5928-51-8 | C7H8O2S | 156.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5928-51-8
Molecular Formula C7H8O2S
Molecular Weight (g/mol) 156.2
MDL Number MFCD00047093
InChI Key MJPVYTKZYZPIQA-UHFFFAOYSA-N
Synonym 3-2-thienyl propanoic acid, 3-2-thienyl propionic acid, 2-thiophenepropanoic acid, 3-thiophen-2-yl propanoic acid, 2-thiophenepropionic acid, maybridge3_005592, 3-2-thienyl propionic pound inverted question markacid, 3-thiophen-2-yl-propionic acid, thiolpropionic acid, pubchem23928
PubChem CID 703169
IUPAC Name 3-thiophen-2-ylpropanoic acid
SMILES C1=CSC(=C1)CCC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 45°C to 49°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in ethanol, acetone
Formula Weight 156.2
Percent Purity 98%
Physical Form Crystals or Needles
Chemical Name or Material 3-(2-Thienyl)propanoic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
TSCA TSCA

RUO – Research Use Only