missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Naphthyl)-D-alanine, 95%

Catalog No. AAH6353214
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6353214 25 g
AAH6353203 1 g
AAH6353206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6353214 Supplier Thermo Scientific Chemicals Supplier No. H6353214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 76985-09-6 | C13H13NO2 | 215.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 76985-09-6
Molecular Formula C13H13NO2
Molecular Weight (g/mol) 215.25
MDL Number MFCD00038546
InChI Key JPZXHKDZASGCLU-GFCCVEGCSA-N
Synonym 3-2-naphthyl-d-alanine, h-d-2-nal-oh, r-2-amino-3-naphthalen-2-yl propanoic acid, r-2-amino-3-naphthalene-2-yl-propionic acid, d-2-naphthylalanine, d-3-2-naphthyl-alanine, unii-571v312ymy, 2-d-naphthylalanine, 2r-2-amino-3-2-naphthyl propanoic acid, 2r-2-amino-3-naphthalen-2-ylpropanoic acid
PubChem CID 2733661
IUPAC Name (2R)-2-amino-3-naphthalen-2-ylpropanoic acid
SMILES [NH3+][C@H](CC1=CC=C2C=CC=CC2=C1)C([O-])=O

Specifications

Color White
Beilstein 4352384
Quantity 25 g
Formula Weight 215.25
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-(2-Naphthyl)-D-alanine
EINECSNumber 616-416-6
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only