missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methyl-thiazol-4-yl)-benzaldehyde, 97%, Thermo Scientific™

Catalog No. CC19404CB
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC19404CB 250mg
CC19404DA 1g
CC19404DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC19404CB Supplier Thermo Scientific Chemicals Supplier No. CC19404CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 850375-05-2
Molecular Formula C11H9NOS
Molecular Weight (g/mol) 203.26
MDL Number MFCD02681946
InChI Key FQQPVEKQPGDYPD-UHFFFAOYSA-N
Synonym 3-2-methyl-thiazol-4-yl-benzaldehyde, 3-2-methyl-1,3-thiazol-4-yl benzaldehyde, 3-2-methylthiazol-4-yl benzaldehyde, 4-3-formylphenyl-2-methyl-1,3-thiazole, 3-2-methyl-4-thiazolyl benzaldehyde, benzaldehyde,3-2-methyl-4-thiazolyl, benzaldehyde, 3-2-methyl-4-thiazolyl
PubChem CID 7127782
IUPAC Name 3-(2-methyl-1,3-thiazol-4-yl)benzaldehyde
SMILES CC1=NC(=CS1)C1=CC(C=O)=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 203.26
Percent Purity 97%
Chemical Name or Material 3-(2-Methyl-thiazol-4-yl)-benzaldehyde
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.