missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Furyl)benzoic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 35461-99-5
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.182
MDL Number MFCD02690979
InChI Key RQVVFGRDMHDHNI-UHFFFAOYSA-N
Synonym 3-furan-2-yl benzoic acid, 3-2-furyl benzoic acid, 3-fur-2-ylbenzoic acid, 3-furan-2-yl-benzoic acid, m-2-furyl benzoic acid, benzoic acid, 3-2-furanyl
PubChem CID 5037933
IUPAC Name 3-(furan-2-yl)benzoic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C2=CC=CO2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC46101CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC46101DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

35461-99-5
188.182
RQVVFGRDMHDHNI-UHFFFAOYSA-N
5037933
C1=CC(=CC(=C1)C(=O)O)C2=CC=CO2
C11H8O3
MFCD02690979
3-furan-2-yl benzoic acid, 3-2-furyl benzoic acid, 3-fur-2-ylbenzoic acid, 3-furan-2-yl-benzoic acid, m-2-furyl benzoic acid, benzoic acid, 3-2-furanyl
3-(furan-2-yl)benzoic acid

Specifications

35461-99-5
100.0
C11H8O3
3-furan-2-yl benzoic acid, 3-2-furyl benzoic acid, 3-fur-2-ylbenzoic acid, 3-furan-2-yl-benzoic acid, m-2-furyl benzoic acid, benzoic acid, 3-2-furanyl
C1=CC(=CC(=C1)C(=O)O)C2=CC=CO2
188.182
188.18
3-(2-Furyl)benzoic acid
96.5
97%
MFCD02690979
RQVVFGRDMHDHNI-UHFFFAOYSA-N
3-(furan-2-yl)benzoic acid
5037933
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents