missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Furyl)acrylic acid, 99%

Catalog No. AAB2165330
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2165330 250 g
AAB2165314 25 g
AAB2165322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2165330 Supplier Thermo Scientific Chemicals Supplier No. B2165330
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 539-47-9 | C7H6O3 | 138.12 g/mol

3-(2-Furyl)acrylic acid is a reactant used in the preparation of heteroaryl-substituted bis-trifluoromethyl carbinols and trifluoroacetophenone derivatives malonyl-CoA decarboxylase (MCD) inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(2-Furyl)acrylic acid is a reactant used in the preparation of heteroaryl-substituted bis-trifluoromethyl carbinols and trifluoroacetophenone derivatives malonyl-CoA decarboxylase (MCD) inhibitors.

Solubility
Soluble in dichloromethane and diethyl ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 539-47-9
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00003257,MFCD11656544
InChI Key ZCJLOOJRNPHKAV-ONEGZZNKSA-N
Synonym 3-2-furyl acrylic acid, e-3-furan-2-yl acrylic acid, 2-furanacrylic acid, 3-furan-2-yl acrylic acid, furanacrylic acid, 2-furalacetic acid, furacrylic acid, furylacrylic acid, 2e-3-furan-2-yl prop-2-enoic acid, 2-propenoic acid, 3-2-furanyl
PubChem CID 643402
SMILES OC(=O)\C=C\C1=CC=CO1

Specifications

Melting Point 140°C to 144°C
Boiling Point 286°C
Quantity 250 g
Beilstein 81046
Merck Index 14,4297
Solubility Information Soluble in dichloromethane and diethyl ether.
IUPAC Name (E)-3-(furan-2-yl)prop-2-enoic acid
Formula Weight 138.12
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3-(2-Furyl)acrylic acid
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement

H315-H319-H335

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.
Precautionary Statement GHS P Statement

P261-P280-P305+P351+P338-P304+P340-P405-P501a

Avoid breathing dust/fume/gas/mist/vapors/spray.

Wear protective gloves/protective clothing/eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Store locked up.

Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 208-718-8
RTECSNumber LT8560000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only