missing translation for 'onlineSavingsMsg'
Learn More

[3-(2-Furyl)-1-methyl-1H-pyrazol-5-yl]methanol 97+%, Thermo Scientific™

Chemical Identifiers

CAS 886851-33-8
Molecular Formula C9H10N2O2
Molecular Weight (g/mol) 178.191
InChI Key BYXYWFXMSJTOKB-UHFFFAOYSA-N
Synonym 3-2-furyl-1-methyl-1h-pyrazol-5-yl methanol, 5-furan-2-yl-2-methylpyrazol-3-yl methanol, 3-2-furyl-1-methylpyrazol-5-yl methan-1-ol, 3-furan-2-yl-1-methyl-1h-pyrazol-5-yl methanol
PubChem CID 18525804
IUPAC Name [5-(furan-2-yl)-2-methylpyrazol-3-yl]methanol
SMILES CN1C(=CC(=N1)C2=CC=CO2)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
CC47009CB
View Documents Promotion Details
N/A 250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC47009DA
View Documents Promotion Details
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

886851-33-8
178.191
3-2-furyl-1-methyl-1h-pyrazol-5-yl methanol, 5-furan-2-yl-2-methylpyrazol-3-yl methanol, 3-2-furyl-1-methylpyrazol-5-yl methan-1-ol, 3-furan-2-yl-1-methyl-1h-pyrazol-5-yl methanol
[5-(furan-2-yl)-2-methylpyrazol-3-yl]methanol
C9H10N2O2
BYXYWFXMSJTOKB-UHFFFAOYSA-N
18525804
CN1C(=CC(=N1)C2=CC=CO2)CO

Specifications

886851-33-8
Amber Glass Bottle
3-2-furyl-1-methyl-1h-pyrazol-5-yl methanol, 5-furan-2-yl-2-methylpyrazol-3-yl methanol, 3-2-furyl-1-methylpyrazol-5-yl methan-1-ol, 3-furan-2-yl-1-methyl-1h-pyrazol-5-yl methanol
CN1C(=CC(=N1)C2=CC=CO2)CO
178.191
178.19
[3-(2-Furyl)-1-methyl-1H-pyrazol-5-yl]methanol
97%
C9H10N2O2
BYXYWFXMSJTOKB-UHFFFAOYSA-N
[5-(furan-2-yl)-2-methylpyrazol-3-yl]methanol
18525804
97%
Videos
SDS
Documents

Documents