missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2-Bromophenyl)propionic Acid 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 99°C |
| Color | Yellow |
| Boiling Point | 186°C |
| Quantity | 5 g |
| Formula Weight | 229.07 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-(2-Bromophenyl)propionic Acid |
Sécurité et manipulation
| TSCA | No |