Learn More
3-(2-Aminoethyl)-1-Boc-azetidine, 95%
CAS: 898271-20-0 | C10H20N2O2 | 200.282 g/mol
Supplier: Thermo Scientific Chemicals H60469MD
| Quantity | 250 mg |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 898271-20-0 | |
| 200.282 | |
| XQNQFOKZUQWWDO-UHFFFAOYSA-N | |
| 44630603 | |
| CC(C)(C)OC(=O)N1CC(C1)CCN |
| C10H20N2O2 | |
| MFCD09878606 | |
| 1-boc-3-aminoethylazetidine, tert-butyl 3-2-aminoethyl azetidine-1-carboxylate, 1-boc-3-aminoethyl azetidine, 1-n-boc-3-aminoethyl azetidine, 1-boc-3-2-aminoethyl azetidine, 3-2-aminoethyl-1-boc-azetidine, 1-azetidinecarboxylicacid, 3-2-aminoethyl-, 1,1-dimethylethyl ester, 1-azetidinecarboxylic acid, 3-2-aminoethyl-, 1,1-dimethylethyl ester, tert-butyl 3-2-aminoethyl-1-azetidinecarboxylate | |
| tert-butyl 3-(2-aminoethyl)azetidine-1-carboxylate |
Specifications
| 3-(2-Aminoethyl)-1-Boc-azetidine | |
| C10H20N2O2 | |
| 1-boc-3-aminoethylazetidine, tert-butyl 3-2-aminoethyl azetidine-1-carboxylate, 1-boc-3-aminoethyl azetidine, 1-n-boc-3-aminoethyl azetidine, 1-boc-3-2-aminoethyl azetidine, 3-2-aminoethyl-1-boc-azetidine, 1-azetidinecarboxylicacid, 3-2-aminoethyl-, 1,1-dimethylethyl ester, 1-azetidinecarboxylic acid, 3-2-aminoethyl-, 1,1-dimethylethyl ester, tert-butyl 3-2-aminoethyl-1-azetidinecarboxylate | |
| CC(C)(C)OC(=O)N1CC(C1)CCN | |
| 250 mg | |
| 44630603 | |
| 95% |
| 898271-20-0 | |
| MFCD09878606 | |
| XQNQFOKZUQWWDO-UHFFFAOYSA-N | |
| tert-butyl 3-(2-aminoethyl)azetidine-1-carboxylate | |
| 200.282 | |
| 200.28 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures