missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Pyrrol-1-yl)benzaldehyde, 95+%, Thermo Scientific™

Chemical Identifiers

CAS 129747-77-9
Molecular Formula C11H9NO
Molecular Weight (g/mol) 171.20
MDL Number MFCD03086140
InChI Key PALTUANHIBXQMX-UHFFFAOYSA-N
Synonym 3-1h-pyrrol-1-yl benzaldehyde, 3-pyrrol-1-yl-benzaldehyde, 3-pyrrol-1-yl benzaldehyde, benzaldehyde,3-1h-pyrrol-1-yl, benzaldehyde, 3-1h-pyrrol-1-yl, acmc-20airb, 3-pyrrolylbenzaldehyde, 3-1-pyrrolyl benzaldehyde
PubChem CID 2776528
IUPAC Name 3-pyrrol-1-ylbenzaldehyde
SMILES O=CC1=CC(=CC=C1)N1C=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC21904CB
SDS View Documents
Thermo Scientific Chemicals
CC21904CB
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC21904EA
SDS View Documents
Thermo Scientific Chemicals
CC21904EA
10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

129747-77-9
171.20
PALTUANHIBXQMX-UHFFFAOYSA-N
2776528
O=CC1=CC(=CC=C1)N1C=CC=C1
C11H9NO
MFCD03086140
3-1h-pyrrol-1-yl benzaldehyde, 3-pyrrol-1-yl-benzaldehyde, 3-pyrrol-1-yl benzaldehyde, benzaldehyde,3-1h-pyrrol-1-yl, benzaldehyde, 3-1h-pyrrol-1-yl, acmc-20airb, 3-pyrrolylbenzaldehyde, 3-1-pyrrolyl benzaldehyde
3-pyrrol-1-ylbenzaldehyde

Specifications

129747-77-9
C11H9NO
250mg
PALTUANHIBXQMX-UHFFFAOYSA-N
3-pyrrol-1-ylbenzaldehyde
2776528
95%
95%
MFCD03086140
3-1h-pyrrol-1-yl benzaldehyde, 3-pyrrol-1-yl-benzaldehyde, 3-pyrrol-1-yl benzaldehyde, benzaldehyde,3-1h-pyrrol-1-yl, benzaldehyde, 3-1h-pyrrol-1-yl, acmc-20airb, 3-pyrrolylbenzaldehyde, 3-1-pyrrolyl benzaldehyde
O=CC1=CC(=CC=C1)N1C=CC=C1
171.20
171.2
3-(1H-Pyrrol-1-yl)benzaldehyde
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'