missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Imidazol-1-ylmethyl)benzaldehyde, ≥97%, Thermo Scientific™

Numéro de catalogue. p-3175359
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC43204CB 250mg
CC43204DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC43204CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC43204CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 102432-05-3
Formule moléculaire C11H10N2O
Poids moléculaire (g/mol) 186.214
Numéro MDL MFCD08271917
Clé InChI JMKQKKQFLYPQDD-UHFFFAOYSA-N
Synonyme 3-1h-imidazol-1-ylmethyl benzaldehyde, benzaldehyde,3-1h-imidazol-1-ylmethyl, 3-imidazol-1-ylmethyl benzaldehyde, 3-1h-imidazol-1-yl methyl benzaldehyde, benzaldehyde, 3-1h-imidazol-1-ylmethyl, acmc-1c5ro, 3-imidazolylmethyl benzaldehyde
PubChem CID 10397537
Nom de l’IUPAC 3-(imidazol-1-ylmethyl)benzaldehyde
SOURIRES C1=CC(=CC(=C1)CN2C=CN=C2)C=O

Spécifications

Nom ou substance chimique 3-(1H-imidazol-1-ylmethyl)benzaldehyde
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber glass bottle
Quantité 250mg
Poids de formule 186.21
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.