missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1h-1,2,4-Triazol-1-ylmethyl)benzaldehyde, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 876316-30-2 |
|---|---|
| Molecular Formula | C10H9N3O |
| Molecular Weight (g/mol) | 187.202 |
| InChI Key | IBVURYHFXBPNCW-UHFFFAOYSA-N |
| Synonym | 3-1h-1,2,4-triazol-1-ylmethyl benzaldehyde, benzaldehyde,3-1h-1,2,4-triazol-1-ylmethyl, 3-1,2,4-triazol-1-ylmethyl benzaldehyde, 3-1h-1,2,4-triazol-1-yl methyl benzaldehyde, 3-1,2,4-triazolylmethyl benzaldehyde |
| PubChem CID | 22360585 |
| IUPAC Name | 3-(1,2,4-triazol-1-ylmethyl)benzaldehyde |
| SMILES | C1=CC(=CC(=C1)CN2C=NC=N2)C=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
CC45404CB
|
Thermo Scientific Chemicals
CC45404CB |
250mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC45404DA
|
Thermo Scientific Chemicals
CC45404DA |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC45404DE
|
Thermo Scientific Chemicals
CC45404DE |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 876316-30-2 | |
| 187.202 | |
| 3-1h-1,2,4-triazol-1-ylmethyl benzaldehyde, benzaldehyde,3-1h-1,2,4-triazol-1-ylmethyl, 3-1,2,4-triazol-1-ylmethyl benzaldehyde, 3-1h-1,2,4-triazol-1-yl methyl benzaldehyde, 3-1,2,4-triazolylmethyl benzaldehyde | |
| 3-(1,2,4-triazol-1-ylmethyl)benzaldehyde |
| C10H9N3O | |
| IBVURYHFXBPNCW-UHFFFAOYSA-N | |
| 22360585 | |
| C1=CC(=CC(=C1)CN2C=NC=N2)C=O |
Specifications
| 876316-30-2 | |
| Amber Glass Bottle | |
| 250mg | |
| IBVURYHFXBPNCW-UHFFFAOYSA-N | |
| 3-(1,2,4-triazol-1-ylmethyl)benzaldehyde | |
| 22360585 | |
| 97% |
| 97% | |
| C10H9N3O | |
| 3-1h-1,2,4-triazol-1-ylmethyl benzaldehyde, benzaldehyde,3-1h-1,2,4-triazol-1-ylmethyl, 3-1,2,4-triazol-1-ylmethyl benzaldehyde, 3-1h-1,2,4-triazol-1-yl methyl benzaldehyde, 3-1,2,4-triazolylmethyl benzaldehyde | |
| C1=CC(=CC(=C1)CN2C=NC=N2)C=O | |
| 187.202 | |
| 187.2 | |
| 3-(1H-1, 2, 4-Triazol-1-ylmethyl)benzaldehyde |