missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[3-(1H-1,2,4-Triazol-1-yl)phényl]méthanol, ≥95%, Thermo Scientific™
Spécifications
Spécifications
| Nom ou substance chimique | [3-(1H-1, 2, 4-triazol-1-yl)phenyl]methanol |
| CAS % minimum | 94.5 |
| CAS % maximal | 100.0 |
| Fourchette de pourcentages d’essai | 95% |
| Emballage | Bouteille en verre ambrée |
| Quantité | 1 g |
| Poids de formule | 175.19 |
| Pourcentage de pureté | 95% |
Sécurité et manipulation
| Hazard Statement | GHS H Statement: Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation. Flammable solid. |
| Precautionary Statement | GHS P Statement: Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Keep container tightly closed. Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |