missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Pyrrolyl)benzoic acid, 97%

Catalog No. AAH3312409
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3312409 10 g
AAH3312403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3312409 Supplier Thermo Scientific Chemicals Supplier No. H3312409

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 61471-45-2 | C11H8NO2 | 186.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 61471-45-2
Molecular Formula C11H8NO2
Molecular Weight (g/mol) 186.19
MDL Number MFCD02656610
InChI Key PODFNQCZFHLJPH-UHFFFAOYSA-M
Synonym 3-1h-pyrrol-1-yl benzoic acid, 3-pyrrol-1-yl-benzoic acid, benzoic acid, 3-1h-pyrrol-1-yl, n-3-carboxy phenylpyrrole, 3-1-pyrrolyl benzoic acid, 3-pyrrol-1-yl benzoic acid, 3-pyrrolylbenzoic acid, 1-3-carboxyphenyl-1h-pyrrole, benzoicacid,3-1h-pyrrol-1-yl, benzoicacid, 3-1h-pyrrol-1-yl
PubChem CID 736537
IUPAC Name 3-pyrrol-1-ylbenzoic acid
SMILES [O-]C(=O)C1=CC=CC(=C1)N1C=CC=C1

Specifications

Melting Point 175°C to 178°C
Quantity 10 g
Formula Weight 187.2
Percent Purity 97%
Chemical Name or Material 3-(1-Pyrrolyl)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only