missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Naphthyl)acrylic Acid 95.0+%, TCI America™

Catalog No. N09725G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N09725G Supplier TCI America Supplier No. N09725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13026-12-5
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00014317,MFCD00014317
InChI Key WPXMLUUYWNHQOR-CMDGGOBGSA-N
Synonym naphthalene-1-acrylic acid, z-3-1-naphthyl acrylic acid, z-3-naphthalen-1-yl-acrylic acid, z-3-naphthalen-1-yl acrylic acid, 2z-3-naphthalen-1-yl prop-2-enoic acid
PubChem CID 12158295
IUPAC Name (2E)-3-(naphthalen-1-yl)prop-2-enoic acid
SMILES OC(=O)\C=C\C1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 212°C
Color White-Yellow
Quantity 5 g
Formula Weight 198.22
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(1-Naphthyl)acrylic Acid