missing translation for 'onlineSavingsMsg'
Learn More

3-(1,3-Dioxolan-2-yl)-2-thiophenecarboxylic acid, 97%, Thermo Scientific™

Numéro de catalogue. CC58801CB
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC58801CB 250mg Amber Glass Bottle
CC58801DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC58801CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC58801CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 934570-44-2
Formule moléculaire C8H8O4S
Poids moléculaire (g/mol) 200.21
Numéro MDL MFCD09702391
Clé InChI ZODQJPSGAHBVHU-UHFFFAOYSA-N
Synonyme 3-1,3-dioxolan-2-yl thiophene-2-carboxylic acid, 3-1,3-dioxolan-2-yl-2-thiophenecarboxylic acid, 2-2-carboxythien-3-yl-1,3-dioxolane, 3-1,3-dioxolan-2-yl thiophene-2-carboxylicacid, 2-thiophenecarboxylicacid, 3-1,3-dioxolan-2-yl
PubChem CID 24229593
Nom de l’IUPAC 3-(1,3-dioxolan-2-yl)thiophene-2-carboxylic acid
SOURIRES OC(=O)C1=C(C=CS1)C1OCCO1

Spécifications

Nom ou substance chimique 3-(1, 3-Dioxolan-2-yl)-2-thiophenecarboxylic acid
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 250mg
Poids de formule 200.21
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.