missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2S)-N-[(1S)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-2-pyrrolidinecarboxamide 98.0+%, TCI America™
$225.56 - $786.63
Chemical Identifiers
| CAS | 910110-45-1 |
|---|---|
| Molecular Formula | C23H30N2O2 |
| Molecular Weight (g/mol) | 366.505 |
| MDL Number | MFCD22200506 |
| InChI Key | PWBULUUPVDXEQS-SFTDATJTSA-N |
| Synonym | (S)-N-[(S)-1-Hydroxy-4-methyl-1,1-diphenylpentan-2-yl]pyrrolidine-2-carboxamide, N-[(1S)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-L-prolinamide, Singh′Cs Catalyst |
| PubChem CID | 11954678 |
| IUPAC Name | (2S)-N-[(2S)-1-hydroxy-4-methyl-1,1-diphenylpentan-2-yl]pyrrolidine-2-carboxamide |
| SMILES | CC(C)CC(C(C1=CC=CC=C1)(C2=CC=CC=C2)O)NC(=O)C3CCCN3 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H1407200MG
|
TCI America
H1407200MG |
200 mg |
Each for $225.56
|
|
|||||
|
H14071G
|
TCI America
H14071G |
1 g |
Each for $786.63
|
|
|||||
Chemical Identifiers
| 910110-45-1 | |
| 366.505 | |
| PWBULUUPVDXEQS-SFTDATJTSA-N | |
| 11954678 | |
| CC(C)CC(C(C1=CC=CC=C1)(C2=CC=CC=C2)O)NC(=O)C3CCCN3 |
| C23H30N2O2 | |
| MFCD22200506 | |
| (S)-N-[(S)-1-Hydroxy-4-methyl-1,1-diphenylpentan-2-yl]pyrrolidine-2-carboxamide, N-[(1S)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-L-prolinamide, Singh′Cs Catalyst | |
| (2S)-N-[(2S)-1-hydroxy-4-methyl-1,1-diphenylpentan-2-yl]pyrrolidine-2-carboxamide |
Specifications
| 910110-45-1 | |
| White | |
| MFCD22200506 | |
| (S)-N-[(S)-1-Hydroxy-4-methyl-1,1-diphenylpentan-2-yl]pyrrolidine-2-carboxamide, N-[(1S)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-L-prolinamide, Singh′Cs Catalyst | |
| CC(C)CC(C(C1=CC=CC=C1)(C2=CC=CC=C2)O)NC(=O)C3CCCN3 | |
| 366.505 | |
| 366.51 | |
| Crystalline Powder |
| 193°C | |
| C23H30N2O2 | |
| 200 mg | |
| PWBULUUPVDXEQS-SFTDATJTSA-N | |
| (2S)-N-[(2S)-1-hydroxy-4-methyl-1,1-diphenylpentan-2-yl]pyrrolidine-2-carboxamide | |
| 11954678 | |
| ≥98.0% (HPLC,T) | |
| (2S)-N-[(1S)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-2-pyrrolidinecarboxamide |
Safety and Handling
TSCA : No