missing translation for 'onlineSavingsMsg'
Learn More

(2S)-N-[(1S)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-2-pyrrolidinecarboxamide 98.0+%, TCI America™

Supplier:  TCI America H14071G

 View more versions of this product

Catalog No. H14071G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

(2S)-N-[(1S)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-2-pyrrolidinecarboxamide
193°C
C23H30N2O2
1 g
PWBULUUPVDXEQS-SFTDATJTSA-N
(2S)-N-[(2S)-1-hydroxy-4-methyl-1,1-diphenylpentan-2-yl]pyrrolidine-2-carboxamide
11954678
≥98.0% (HPLC,T)
910110-45-1
White
MFCD22200506
(S)-N-[(S)-1-Hydroxy-4-methyl-1,1-diphenylpentan-2-yl]pyrrolidine-2-carboxamide, N-[(1S)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-L-prolinamide, Singh′Cs Catalyst
CC(C)CC(C(C1=CC=CC=C1)(C2=CC=CC=C2)O)NC(=O)C3CCCN3
366.505
366.51
Crystalline Powder

Chemical Identifiers

910110-45-1
366.505
PWBULUUPVDXEQS-SFTDATJTSA-N
11954678
CC(C)CC(C(C1=CC=CC=C1)(C2=CC=CC=C2)O)NC(=O)C3CCCN3
C23H30N2O2
MFCD22200506
(S)-N-[(S)-1-Hydroxy-4-methyl-1,1-diphenylpentan-2-yl]pyrrolidine-2-carboxamide, N-[(1S)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-L-prolinamide, Singh′Cs Catalyst
(2S)-N-[(2S)-1-hydroxy-4-methyl-1,1-diphenylpentan-2-yl]pyrrolidine-2-carboxamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions