CAS RN 910110-45-1
CAS RN 910110-45-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(2S)-N-[(1S)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-2-pyrrolidinecarboxamide 98.0+%, TCI America™
CAS: 910110-45-1 Molecular Formula: C23H30N2O2 Molecular Weight (g/mol): 366.505 MDL Number: MFCD22200506 InChI Key: PWBULUUPVDXEQS-SFTDATJTSA-N Synonym: (S)-N-[(S)-1-Hydroxy-4-methyl-1,1-diphenylpentan-2-yl]pyrrolidine-2-carboxamide, N-[(1S)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-L-prolinamide, Singh′Cs Catalyst PubChem CID: 11954678 IUPAC Name: (2S)-N-[(2S)-1-hydroxy-4-methyl-1,1-diphenylpentan-2-yl]pyrrolidine-2-carboxamide SMILES: CC(C)CC(C(C1=CC=CC=C1)(C2=CC=CC=C2)O)NC(=O)C3CCCN3