missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2S,5S)-2,5-Hexanediol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 54°C |
| Color | White-Yellow |
| Boiling Point | 71°C |
| Quantity | 5 g |
| Formula Weight | 118.18 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (2S,5S)-2,5-Hexanediol |
Safety and Handling
| EINECSNumber | (2)-0240 |
| TSCA | No |