missing translation for 'onlineSavingsMsg'
Learn More

(2S,4R)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic Acid 96.0+%, TCI America™

Catalog No. p-7138105
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3177200MG 200 mg
B31771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3177200MG Supplier TCI America Supplier No. B3177200MG

Chemical Identifiers

CAS 203866-14-2
Molecular Formula C10H16FNO4
Molecular Weight (g/mol) 233.239
MDL Number MFCD06796085
InChI Key YGWZXQOYEBWUTH-RQJHMYQMSA-N
Synonym n-boc-trans-4-fluoro-l-proline, 2s,4r-1-tert-butoxycarbonyl-4-fluoropyrrolidine-2-carboxylic acid, boc-trans-4-fluoro-l-proline, n-tert-butoxycarbonyl-2s,4r-4-fluoroproline, 2s,4r-1-boc-4-fluoro-2-pyrrolidinecarboxylic acid, boc-trans-4-fluoro-pro-oh, 2s,4r-1-tert-butoxycarbonyl-4-fluoro-2-pyrrolidinecarboxylic acid, 2s,4r-1-tert-butoxy carbonyl-4-fluoropyrrolidine-2-carboxylic acid, trans-4-fluoro-l-proline, n-boc protected, 2s,4r-1-tert-butoxycarbonyl-4-fluoropyrrolidine-2-carboyxlic acid
PubChem CID 12043048
IUPAC Name (2S,4R)-4-fluoro-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(CC1C(=O)O)F

Specifications

Melting Point 116°C
Color White-Yellow
Quantity 200 mg
Formula Weight 233.24
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (2S,4R)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic Acid