missing translation for 'onlineSavingsMsg'
Learn More

(2S,3S)-3-(tert-Butoxycarbonylamino)-1,2-epoxy-4-phenylbutane 98.0+%, TCI America™

Catalog No. E08935G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08935G 5 g
E08931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08935G Supplier TCI America Supplier No. E08935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98737-29-2
Molecular Formula C15H21NO3
Molecular Weight (g/mol) 263.337
MDL Number MFCD02258997
InChI Key NVPOUMXZERMIJK-QWHCGFSZSA-N
Synonym 2s,3s-1,2-epoxy-3-boc-amino-4-phenylbutane, 2s,3s-n-t-boc-3-amino-1,2-epoxy-4-phenylbutane, tert-butyl s-1-s-oxiran-2-yl-2-phenylethyl carbamate, 2s,3s-1,2-epoxy-3-tert-butoxycarbonylamino-4-phenylbutane, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl s-r*,r*---1-oxiranyl-2-phenylethyl carbamate, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenyl-ethyl carbamate, 2s,3s---3-t-boc-amino-1,2-epoxy-4-phenylbutane, ksc523o6b
PubChem CID 9903372
IUPAC Name tert-butyl N-[(1S)-1-[(2S)-oxiran-2-yl]-2-phenylethyl]carbamate
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C2CO2

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 5 g
UN Number 3077
Formula Weight 263.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (2S,3S)-3-(tert-Butoxycarbonylamino)-1,2-epoxy-4-phenylbutane
EINECSNumber (5)-6533
TSCA No